Assay Details
| Gene Symbol: PTCHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746624.3 | XP_002746670.2 | 2 - 3 | 1082 | 67 | |
| XM_002746625.3 | XP_002746671.1 | 2 - 3 | 908 | 67 | ||
| XM_008994552.2 | XP_008992800.1 | 3 - 4 | 1239 | 67 | ||
| XM_017971145.1 | XP_017826634.1 | 3 - 4 | 1065 | 67 | ||
More Information
Set Membership: |
|

PTCHD4
Genomic Map