Assay Details
| Gene Symbol: ZFP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755308.4 | XP_002755354.2 | 3 - 4 | 799 | 93 | |
| XM_009008164.2 | XP_009006412.1 | 3 - 4 | 799 | 93 | ||
| XM_009008165.2 | XP_009006413.1 | 3 - 4 | 799 | 93 | ||
More Information
Set Membership: |
|

ZFP91
Genomic Map