Assay Details
Gene Symbol: CYP26C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756409.3 | XP_002756455.1 | 3 - 4 | 1112 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
