Assay Details
| Gene Symbol: STAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759723.2 | XP_002759769.1 | 2 - 3 | 547 | 61 | |
| XM_008984000.2 | XP_008982248.1 | 2 - 3 | 547 | 61 | ||
More Information
Set Membership: |
|

STAC
Genomic Map