Assay Details
| Gene Symbol: EMR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987432.2 | XP_008985680.1 | 14 - 15 | 1697 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMR3| Gene Symbol: EMR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987432.2 | XP_008985680.1 | 14 - 15 | 1697 | 107 | |
Set Membership: |
|