Assay Details
| Gene Symbol: EREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993179.2 | XP_008991427.1 | 4 - 5 | 823 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EREG| Gene Symbol: EREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993179.2 | XP_008991427.1 | 4 - 5 | 823 | 121 | |
Set Membership: |
|