Assay Details
| Gene Symbol: GPAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964058.1 | XP_017819547.1 | 2 - 3 | 236 | 88 | |
| XM_017964059.1 | XP_017819548.1 | 2 - 3 | 177 | 88 | ||
More Information
Set Membership: |
|

GPAT2
Genomic Map