Assay Details
| Gene Symbol: FOLH1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754698.2 | XP_002754744.1 | 14 - 15 | 1790 | 92 | |
| XM_009007272.2 | XP_009005520.1 | 14 - 15 | 1708 | 92 | ||
| XM_009007273.1 | XP_009005521.1 | 14 - 15 | 1790 | 92 | ||
| XM_017977700.1 | XP_017833189.1 | 15 - 16 | 1750 | 92 | ||
More Information
Set Membership: |
|

FOLH1B
Genomic Map