Assay Details
| Gene Symbol: IRF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755688.3 | XP_002755734.1 | 3 - 4 | 515 | 71 | |
| XM_009008765.2 | XP_009007013.1 | 3 - 4 | 515 | 71 | ||
| XM_009008766.2 | XP_009007014.1 | 3 - 4 | 515 | 71 | ||
More Information
Set Membership: |
|

IRF7
Genomic Map