Assay Details
| Gene Symbol: ACTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762087.3 | XP_002762133.1 | 3 - 4 | 534 | 65 | |
| XM_008987951.2 | XP_008986199.1 | 3 - 4 | 534 | 65 | ||
| XM_008987952.2 | XP_008986200.1 | 3 - 4 | 534 | 65 | ||
| XM_008987953.2 | XP_008986201.1 | 3 - 4 | 534 | 65 | ||
| XM_008987955.2 | XP_008986203.1 | 3 - 4 | 534 | 65 | ||
| XM_008987956.2 | XP_008986204.1 | 3 - 4 | 534 | 65 | ||
More Information
Set Membership: |
|

ACTN4
Genomic Map