Assay Details
| Gene Symbol: VEGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762646.3 | XP_002762692.1 | 3 - 4 | 1256 | 94 | |
| XM_008988985.2 | XP_008987233.1 | 3 - 4 | 1253 | 94 | ||
| XM_008988986.2 | XP_008987234.1 | 3 - 4 | 1256 | 94 | ||
More Information
Set Membership: |
|

VEGFD
Genomic Map