Assay Details
| Gene Symbol: CCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732986.3 | XP_003733034.2 | 1 - 2 | 525 | 77 | |
| XM_017972535.1 | XP_017828024.1 | 1 - 2 | 109 | 77 | ||
More Information
Set Membership: |
|

CCL8
Genomic Map