Assay Details
| Gene Symbol: LOC100401798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979614.2 | XP_008977862.1 | 17 - 18 | 2344 | 78 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100401798| Gene Symbol: LOC100401798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979614.2 | XP_008977862.1 | 17 - 18 | 2344 | 78 | |