Assay Details
| Gene Symbol: ANKRD30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973949.1 | XP_017829438.1 | 32 - 33 | 3972 | 82 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD30A| Gene Symbol: ANKRD30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973949.1 | XP_017829438.1 | 32 - 33 | 3972 | 82 | |