Assay Details
| Gene Symbol: C1H9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974378.1 | XP_017829867.1 | 2 - 3 | 1321 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1H9orf131| Gene Symbol: C1H9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974378.1 | XP_017829867.1 | 2 - 3 | 1321 | 87 | |
Set Membership: |
|