Assay Details
Gene Symbol: LRIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002751209.3 | XP_002751255.1 | 1 - 2 | 291 | 91 | |
XM_009001862.2 | XP_009000110.1 | 1 - 2 | 291 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
