Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756144.4 | XP_002756190.2 | 2 - 3 | 1662 | 86 | |
XM_009009599.2 | XP_009007847.1 | 3 - 4 | 1655 | 86 | ||
XM_009009601.2 | XP_009007849.1 | 3 - 4 | 1655 | 86 | ||
XM_009009602.2 | XP_009007850.1 | 3 - 4 | 1655 | 86 | ||
XM_017978940.1 | XP_017834429.1 | 4 - 5 | 1731 | 86 | ||
XM_017978941.1 | XP_017834430.1 | 3 - 4 | 1655 | 86 | ||
XM_017978942.1 | XP_017834431.1 | 3 - 4 | 1655 | 86 |
