Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756144.4 | XP_002756190.2 | 3 - 4 | 1774 | 106 | |
XM_009009599.2 | XP_009007847.1 | 4 - 5 | 1767 | 106 | ||
XM_009009601.2 | XP_009007849.1 | 4 - 5 | 1767 | 106 | ||
XM_009009602.2 | XP_009007850.1 | 4 - 5 | 1767 | 106 | ||
XM_009009603.2 | XP_009007851.1 | 3 - 4 | 243 | 106 | ||
XM_017978940.1 | XP_017834429.1 | 5 - 6 | 1843 | 106 | ||
XM_017978941.1 | XP_017834430.1 | 4 - 5 | 1767 | 106 | ||
XM_017978942.1 | XP_017834431.1 | 4 - 5 | 1767 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009009603.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
