Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756144.4 | XP_002756190.2 | 6 - 7 | 2308 | 85 | |
XM_009009599.2 | XP_009007847.1 | 7 - 8 | 2301 | 85 | ||
XM_009009601.2 | XP_009007849.1 | 7 - 8 | 2301 | 85 | ||
XM_009009603.2 | XP_009007851.1 | 6 - 7 | 777 | 85 | ||
XM_017978940.1 | XP_017834429.1 | 8 - 9 | 2377 | 85 | ||
XM_017978941.1 | XP_017834430.1 | 7 - 8 | 2301 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
