Assay Details
| Gene Symbol: MAFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743769.3 | XP_002743815.1 | 3 | 894 | 107 | |
| XM_008979066.2 | XP_008977314.1 | 3 | 898 | 107 | ||
| XM_008979069.2 | XP_008977317.1 | 3 | 981 | 107 | ||
| XM_008979073.2 | XP_008977321.1 | 3 | 1184 | 107 | ||
| XM_008979078.2 | XP_008977326.1 | 3 | 1100 | 107 | ||
| XM_017963659.1 | XP_017819148.1 | 3 | 756 | 107 | ||
More Information
Set Membership: |
|

MAFF
Genomic Map