Assay Details
| Gene Symbol: CFAP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750729.2 | XP_002750775.1 | 22 - 23 | 3682 | 58 | |
| XM_009001048.1 | XP_008999296.1 | 22 - 23 | 3685 | 58 | ||
| XM_017974620.1 | XP_017830109.1 | 23 - 24 | 3772 | 58 | ||
More Information
Set Membership: |
|

CFAP57
Genomic Map