Assay Details
| Gene Symbol: DDIAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754747.3 | XP_002754793.1 | 6 - 7 | 867 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDIAS| Gene Symbol: DDIAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754747.3 | XP_002754793.1 | 6 - 7 | 867 | 75 | |
Set Membership: |
|