Assay Details
| Gene Symbol: PKMYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755817.3 | XP_002755863.1 | 4 - 5 | 1377 | 72 | |
| XM_009009008.2 | XP_009007256.1 | 3 - 4 | 999 | 72 | ||
| XM_017978657.1 | XP_017834146.1 | 4 - 5 | 1161 | 72 | ||
| XM_017978658.1 | XP_017834147.1 | 4 - 5 | 1250 | 72 | ||
More Information
Set Membership: |
|

PKMYT1
Genomic Map