Assay Details
| Gene Symbol: LOC100394842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997150.2 | XP_008995398.1 | 3 - 4 | 450 | 74 | |
| XM_017972558.1 | XP_017828047.1 | 4 - 5 | 1685 | 74 | ||
| XM_017972559.1 | XP_017828048.1 | 4 - 5 | 1689 | 74 | ||
More Information
Set Membership: |
|

LOC100394842
Genomic Map