Assay Details
| Gene Symbol: LOC100394842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997150.2 | XP_008995398.1 | 5 - 6 | 645 | 92 | |
| XM_017972558.1 | XP_017828047.1 | 6 - 7 | 1880 | 92 | ||
| XM_017972559.1 | XP_017828048.1 | 6 - 7 | 1881 | 92 | ||
More Information
Set Membership: |
|

LOC100394842
Genomic Map