Assay Details
| Gene Symbol: WRAP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000300.2 | XP_008998548.1 | 8 - 9 | 916 | 67 | |
| XM_009000301.2 | XP_008998549.1 | 8 - 9 | 916 | 67 | ||
| XM_017974248.1 | XP_017829737.1 | 7 - 8 | 829 | 67 | ||
| XM_017974249.1 | XP_017829738.1 | 6 - 7 | 646 | 67 | ||
More Information
Set Membership: |
|

WRAP73
Genomic Map