Assay Details
| Gene Symbol: THEGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745809.4 | XP_002745855.2 | 1 - 2 | 583 | 93 | |
| XM_008993279.2 | XP_008991527.1 | 1 - 2 | 583 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

THEGL| Gene Symbol: THEGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745809.4 | XP_002745855.2 | 1 - 2 | 583 | 93 | |
| XM_008993279.2 | XP_008991527.1 | 1 - 2 | 583 | 93 | ||