Assay Details
| Gene Symbol: PLA2G4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002807198.4 | XP_002807244.2 | 19 - 20 | 2588 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G4E| Gene Symbol: PLA2G4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002807198.4 | XP_002807244.2 | 19 - 20 | 2588 | 110 | |
Set Membership: |
|