Assay Details
| Gene Symbol: LOC103787275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967405.1 | XP_017822894.1 | 8 - 9 | 2149 | 57 | |
| XM_017967406.1 | XP_017822895.1 | 7 - 8 | 1868 | 57 | ||
More Information
Set Membership: |
|

LOC103787275
Genomic Map