Assay Details
| Gene Symbol: LOC100400178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017969515.1 | XP_017825004.1 | 6 - 7 | 815 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100400178| Gene Symbol: LOC100400178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017969515.1 | XP_017825004.1 | 6 - 7 | 815 | 134 | |
Set Membership: |
|