Assay Details
| Gene Symbol: TRAPPC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746889.3 | XP_002746935.1 | 1 - 2 | 130 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAPPC3L| Gene Symbol: TRAPPC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746889.3 | XP_002746935.1 | 1 - 2 | 130 | 69 | |
Set Membership: |
|