Assay Details
| Gene Symbol: AGXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749974.3 | XP_002750020.2 | 6 - 7 | 1029 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGXT| Gene Symbol: AGXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749974.3 | XP_002750020.2 | 6 - 7 | 1029 | 61 | |
Set Membership: |
|