Assay Details
| Gene Symbol: NCMAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750429.3 | XP_002750475.1 | 3 - 4 | 235 | 61 | |
| XM_017974407.1 | XP_017829896.1 | 3 - 4 | 647 | 61 | ||
| XM_017974408.1 | XP_017829897.1 | 3 - 4 | 354 | 61 | ||
More Information
Set Membership: |
|

NCMAP
Genomic Map