Assay Details
| Gene Symbol: STK32B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746002.2 | XP_002746048.1 | 7 - 8 | 1020 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STK32B| Gene Symbol: STK32B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746002.2 | XP_002746048.1 | 7 - 8 | 1020 | 61 | |
Set Membership: |
|