Assay Details
| Gene Symbol: SPNS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747846.2 | XP_002747892.1 | 4 - 5 | 595 | 64 | |
| XM_017972228.1 | XP_017827717.1 | 3 - 4 | 464 | 64 | ||
More Information
Set Membership: |
|

SPNS3
Genomic Map