Assay Details
| Gene Symbol: CD79B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748119.2 | XP_002748165.1 | 2 - 3 | 201 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD79B| Gene Symbol: CD79B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748119.2 | XP_002748165.1 | 2 - 3 | 201 | 65 | |
Set Membership: |
|