Assay Details
| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753337.3 | XP_002753383.1 | 1 - 2 | 189 | 84 | |
| XM_009005221.2 | XP_009003469.1 | 1 - 2 | 189 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

REC114| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753337.3 | XP_002753383.1 | 1 - 2 | 189 | 84 | |
| XM_009005221.2 | XP_009003469.1 | 1 - 2 | 189 | 84 | ||