Assay Details
| Gene Symbol: NT5C1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981100.2 | XP_008979348.1 | 6 - 7 | 1128 | 89 | |
| XM_008981102.2 | XP_008979350.1 | 5 - 6 | 948 | 89 | ||
| XM_017964525.1 | XP_017820014.1 | 6 - 7 | 1179 | 89 | ||
| XM_017964526.1 | XP_017820015.1 | 6 - 7 | 1134 | 89 | ||
| XM_017964527.1 | XP_017820016.1 | 6 - 7 | 1077 | 89 | ||
More Information
Set Membership: |
|

NT5C1B
Genomic Map