Assay Details
| Gene Symbol: RSPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995618.1 | XP_008993866.1 | 1 - 2 | 157 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSPO4| Gene Symbol: RSPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995618.1 | XP_008993866.1 | 1 - 2 | 157 | 68 | |
Set Membership: |
|