Assay Details
| Gene Symbol: SLC23A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744217.3 | XP_002744263.1 | 13 - 14 | 1980 | 68 | |
| XM_008985672.2 | XP_008983920.1 | 13 - 14 | 1996 | 68 | ||
| XM_008985678.2 | XP_008983926.1 | 13 - 14 | 1995 | 68 | ||
| XM_008985684.2 | XP_008983932.1 | 13 - 14 | 1997 | 68 | ||
| XM_008985703.2 | XP_008983951.1 | 13 - 14 | 1979 | 68 | ||
More Information
Set Membership: |
|

SLC23A1
Genomic Map