Assay Details
| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748040.3 | XP_002748086.1 | 2 - 3 | 275 | 79 | |
| XM_017973109.1 | XP_017828598.1 | 2 - 3 | 275 | 79 | ||
| XM_017973110.1 | XP_017828599.1 | 2 - 3 | 266 | 79 | ||
More Information
Set Membership: |
|

PPY
Genomic Map