Assay Details
| Gene Symbol: GUCA1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758826.3 | XP_002758872.1 | 1 - 2 | 317 | 99 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCA1C| Gene Symbol: GUCA1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758826.3 | XP_002758872.1 | 1 - 2 | 317 | 99 | |