Assay Details
| Gene Symbol: FMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760374.3 | XP_002760420.1 | 7 - 8 | 1335 | 57 | |
| XM_017966094.1 | XP_017821583.1 | 4 - 5 | 902 | 57 | ||
More Information
Set Membership: |
|

FMO2
Genomic Map