Assay Details
| Gene Symbol: GABRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991013.2 | XP_008989261.1 | 6 - 7 | 771 | 68 | |
| XM_008991019.2 | XP_008989267.1 | 6 - 7 | 842 | 68 | ||
| XM_017969679.1 | XP_017825168.1 | 6 - 7 | 773 | 68 | ||
More Information
Set Membership: |
|

GABRP
Genomic Map