Assay Details
Gene Symbol: ARL5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017971733.1 | XP_017827222.1 | 4 - 5 | 362 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
