Assay Details
| Gene Symbol: ATG9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975513.1 | XP_017831002.1 | 5 - 6 | 1437 | 50 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATG9B| Gene Symbol: ATG9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975513.1 | XP_017831002.1 | 5 - 6 | 1437 | 50 | |
Set Membership: |
|