Assay Details
| Gene Symbol: CALCOCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748465.4 | XP_002748511.2 | 5 - 6 | 649 | 93 | |
| XM_008997418.2 | XP_008995666.1 | 5 - 6 | 649 | 93 | ||
| XM_008997419.2 | XP_008995667.1 | 5 - 6 | 649 | 93 | ||
| XM_017972692.1 | XP_017828181.1 | 5 - 6 | 649 | 93 | ||
More Information
Set Membership: |
|

CALCOCO2
Genomic Map