Assay Details
| Gene Symbol: KLRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752334.3 | XP_002752380.1 | 2 - 3 | 394 | 63 | |
| XM_017975945.1 | XP_017831434.1 | 2 - 3 | 403 | 63 | ||
More Information
Set Membership: |
|

KLRG1
Genomic Map