Assay Details
| Gene Symbol: BPNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985189.2 | XP_008983437.1 | 682 | 60 | ||
| XM_017966475.1 | XP_017821964.1 | 675 | 60 | |||
| XM_017966476.1 | XP_017821965.1 | 738 | 60 | |||
| XM_017966477.1 | XP_017821966.1 | 728 | 60 | |||
More Information
Set Membership: |
|

BPNT1
Genomic Map