Assay Details
| Gene Symbol: MAGEA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008990052.2 | XP_008988300.1 | 5 - 6 | 382 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAGEA11| Gene Symbol: MAGEA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008990052.2 | XP_008988300.1 | 5 - 6 | 382 | 67 | |
Set Membership: |
|